EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Interaction of a sulfhydryl analogue of vasoactive intestinal peptide (VIP) with murine lymphocytes.
PMID 8525988
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V4380
[D-p-Cl-Phe6, Leu17]-Vasoactive Intestinal Peptide human, porcine, rat, ≥97% (HPLC)
C148H239ClN44O42