EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
Recessively inherited L-DOPA-responsive dystonia caused by a point mutation (Q381K) in the tyrosine hydroxylase gene.
PMID 8528210
Abstract
Tyrosine hydroxylase (TH) catalyzes the conversion of L-tyrosine to L-dihydroxyphenylalanine (L-DOPA), the rate-limiting step in the biosynthesis of dopamine. Recently, we described a point mutation in hTH (Q381K) in a family of two siblings suffering from progressive L-DOPA-responsive dystonia (DRD), representing the first reported mutation in this gene. We here describe the cloning, expression and steady-state kinetic properties of the recombinant mutant enzyme. When expressed by a coupled in vitro transcription-translation system and in E. coli, the mutant enzyme represents a kinetic variant form, with a reduced affinity for L-tyrosine. The 'residual activity' of about 15% of the corresponding wild-type hTH (isoform hTH1), at substrate concentrations prevailing in vivo, is compatible with the clinical phenotype of the two Q381K homozygote patients carrying this recessively inherited mutation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93829
L-Tyrosine, BioUltra, ≥99.0% (NT)
C9H11NO3