EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Rapid purification of fluorescent dye-labeled products in a 96-well format for high-throughput automated DNA sequencing.
PMID 8532542
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G75120 Sephadex® G-75, Medium