EMAIL THIS PAGE TO A FRIEND
Bioorganicheskaia khimiia
[Synthesis and biological properties of pyrimidine 2'-deoxynucleosides modified by a residue of quinaldic acid].
PMID 8540903
Abstract
O-Quinaldoyl derivatives of thymidine, 2'-deoxyuridine, and 5-trimethylsilyl-2'-deoxyuridine were synthesized. 5'-Deoxy-5'-(quinoline-2-carbonylamino)- and 5'-deoxy-5'-[(quinoline-2-carbonylamino)butyroylamino]thymidine were obtained by the reaction of 5'-amino-5'-deoxythymidine with pentafluorophenyl ester of quinaldic acid, or with 4-(quinoline-2-carbonylamino)butyric acid. Antiproliferative properties in respect to CaOv cells in vitro were found in most of the synthesized quinaldoyl derivatives of nucleosides (CE50 approximately 10(-5) M). 3'-O-Quinaldoylthymidine exhibited an antitumor activity in vivo. The interaction of 3'- and 5'-O-quinaldoyl- as well as 3',5'-di-O-quinaldoylthymidine with DNA was investigated by the method of fluorescent probes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
160660
Quinaldic acid, 98%
C10H7NO2