EMAIL THIS PAGE TO A FRIEND
Acta neurobiologiae experimentalis
Neurotoxic effect of sodium tellurite in the rat temporal lobe.
PMID 8553916
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
400688
Sodium tellurite, −100 mesh, 99%
Na2O3Te