EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Fixed drug eruption due to citiolone.
PMID 8565496
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A16602
DL-N-Acetylhomocysteine thiolactone, 98%
C6H9NO2S