EMAIL THIS PAGE TO A FRIEND
Biochemical Society transactions
Ultra-rapid detection of mRNAs on northern blots with digoxigenin-labelled oligonucleotides and 'CDP-Star', a new chemiluminescence substrate.
PMID 8566394