EMAIL THIS PAGE TO A FRIEND
Biochemical Society transactions
The role of microtubules in apical and basolateral endocytosis in epithelial Madin-Darby canine kidney (MDCK) cells.
PMID 8566409
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
MDCK Cell Line canine, 84121903