EMAIL THIS PAGE TO A FRIEND
Der Hautarzt; Zeitschrift fur Dermatologie, Venerologie, und verwandte Gebiete
[Therapy of psoriasis with fumaderm preparation].
PMID 8567277
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
128422
mono-Ethyl fumarate, 95%
C6H8O4