EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Deprotection of S-acetamidomethyl cysteine-containing peptides by silver trifluoromethanesulfonate avoids the oxidation of methionines.
PMID 8572278
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00320
S-Acetamidomethyl-L-cysteine hydrochloride, ≥99.0% (AT)
C6H12N2O3S · HCl