EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Detection of specific bacterial cells with 2-hydroxy-3-naphthoic acid-2'-phenylanilide phosphate and fast red TR in situ hybridization.
PMID 8572706
Abstract
An in situ hybridization technique with HNPP (2-hydroxy-3-naphthoic acid-2'-phenylanilide phosphate) and Fast Red TR was used to detect specific bacterial cells at the single-cell level. By this technique, the fluorescent signals of target bacterial cells were up to eight times more intense than those of standard fluorescence in situ hybridization with mono-fluorescein isothiocyanate-labeled oligonucleotide probes. This novel HNPP-Fast Red TR whole-cell hybridization technique is available for the identification of small or low-rRNA-content bacterial cells in the natural environment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
368881
Fast Red TR Salt hemi(zinc chloride) salt, Dye content 15 %
C7H6Cl2N2 · 0.5 ZnCl2