EMAIL THIS PAGE TO A FRIEND
Journal of cancer research and clinical oncology
Oral antitumour activity in murine L1210 leukaemia and pharmacological properties of liposome formulations of N4-alkyl derivatives of 1-beta-D-arabinofuranosylcytosine.
PMID 8576276
Abstract
The oral cytostatic activity in L1210 mouse leukaemia of the two new N4-alkyl derivatives of 1-beta-D-arabinofuranosylcytosine (AraC), N4-hexadecyl- and N4-octadecyl-1-beta-D-arabinofuranosylcytosine (NH-AraC, NO-AraC) was investigated. In contrast to AraC, both derivatives were highly cytostatic after oral application as liposome formulations. With treatment schedules of five consecutive dosages or with two applications on days 1 and 4 after intravenous tumour cell inoculation with a total dose of 470-1000 mg/kg NH-AraC or NO-AraC, 70%-100% of the treated animals were cured. The lethal dose in healthy ICR mice after a single intraperitoneal application, corresponding to the LD50, was 524 mg/kg for NO-AraC, whereas NH-AraC proved to be less toxic. The haematological toxicity remained moderate for both drugs with a mild leucopenia and a drop in platelet counts, which recovered 4-6 days after treatment. The erythrocytes were not affected and haemolytic toxicities were absent. As non-haematological toxicities, at high drug concentrations, a pronounced atrophy of the rapidly dividing epithelia of the small intestines and of the white pulp of the spleen were observed. The blood levels of NH-AraC given orally reached values comparable to those after parenteral application of a four-times lower dose of NH-AraC, suggesting a moderate bioavailability. Thus, these two lipophilic derivatives of AraC are compounds with a potential for the oral treatment of malignant diseases.