EMAIL THIS PAGE TO A FRIEND
Journal of accident & emergency medicine
Occupational phosgene poisoning: a case report and review.
PMID 8581252
Abstract
Phosgene is a highly toxic gas to which some workers may be occupationally exposed. This case report demonstrates the possibility of refrigeration workers suffering phosgene poisoning after heating certain chlorinated fluorocarbons ('freons'). The need to suspect phosgene exposure and observe such patients is emphasized, especially in view of the delay in clinical deterioration observed in some patients who subsequently develop adult respiratory distress syndrome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
748684
Phosgene solution, 15 wt. % in toluene
CCl2O