EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Cytolysis of human erythrocytes by a covalent antibody-selenium immunoconjugate.
PMID 8582643
Abstract
A selenium (Se)-containing immunoconjugate of a human anti-erythrocyte membrane antibody (Ab-Se) has been synthesized via oxidation of the carbohydrate moieties of the antibody and covalent coupling with selenocystamine. The isolated Ab-Se immunoconjugate is shown to be more hemolytic than is selenocystamine when expressed on equivalent selenium basis. Native antibody preincubated with the human erythrocytes prevented hemolysis induced by the Ab-Se immunoconjugate. As observed microscopically, the Ab-Se immunoconjugate caused extensive damage to the erythrocyte membrane and lysis. The cytotoxicity of Se toward the human erythrocytes is believed to be caused initially by the localized generation of superoxide (O2.-) within the cell membrane. This is the first demonstration of site-directed immunoselectivity of Se cytotoxicity and demonstrates the potential for a free radical pharmacology based on localized Se-generated O2.-.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0520
Selenocystamine dihydrochloride, powder
C4H12N2Se2 · 2HCl