EMAIL THIS PAGE TO A FRIEND
HNO
[Bismuth gallate in wound treatment].
PMID 8582836
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
15102
Bismuth(III) gallate basic hydrate, ≥98.0% (calc. on dry substance, KT)
C7H5BiO6 · xH2O