EMAIL THIS PAGE TO A FRIEND
The Canadian veterinary journal = La revue veterinaire canadienne
[Esophageal closure and the efficacy of febantel against Muellerius capillaris in goats].
PMID 8590430
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33981
Febantel, VETRANAL®, analytical standard
C20H22N4O6S