EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Synthesis of (1-->4)-beta-D-xylo-oligosaccharides of dp 4-10 by a blockwise approach.
PMID 8590447
Abstract
Dibutyltin oxide-mediated regioselective chloroacetylation of methyl 1-thio-beta-xylobioside, followed by treatment of the product with 4-methylbenzoyl chloride-pyridine, gave methyl 4-O-chloroacetyl-2,3-di-O-(4-methylbenzoyl)-beta-D-xylopyranosyl-(1-->4) -2.3-di - O-(4-methylbenzoyl)-1-thio-beta-D-xylopyranoside (18) in 70% yield. Coupling of 18 with benzyl alcohol afforded the disaccharide benzyl beta-glycoside, which was O-dechloroacetylated to provide methyl 2,3-di-O-(4-methylbenzoyl)-beta-D-xylopyranosyl-(1-->4)-2,3-di-O-(4- methylbenzoyl)-1-thio-beta-D-xylopyranoside (20). A homologous series of (1-->4)-beta-D-xylo-oligosaccharides from the tetra- to the deca-saccharide have been synthesized in a blockwise manner by using 20 as the glycosyl acceptor, 18, methyl 1-thio-beta-xylobioside pentaacetate, and methyl 1-thio-beta-xylotrioside heptaacetate as the glycosyl donors, and a combination of N-iodosuccinimide-silver triflate as the promoter.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34980
Dibutyltin(IV) oxide, purum
C8H18OSn
183083
Dibutyltin(IV) oxide, 98%
C8H18OSn