EMAIL THIS PAGE TO A FRIEND
Doklady Akademii nauk SSSR
[Effect of inhibitors of lipoxygenase oxidation of arachidonic acid and phenofluorazole on rat thymocyte cytoplasmic pH and on the change of this parameter induced by concanavalin A].
PMID 8593766
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36504
Fenazaflor, PESTANAL®, analytical standard
C15H7Cl2F3N2O2