EMAIL THIS PAGE TO A FRIEND
Survey of ophthalmology
Vitreous amyloidosis in familial amyloidotic polyneuropathy. Report of a case with the Val30Met transthyretin mutation.
PMID 8599155
Abstract
We present a clinical pathological review of vitreous amyloidosis in a case of familial amyloidotic polyneuropathy, type I. Vitreous opacification was the first manifestation of disease in the proband, who was successfully treated with vitrectomy. The eyes were obtained at autopsy after the patient died from an unrelated cause, and the histopathology is presented here. Analysis of DNA from the pathology specimen revealed the most commonly reported transthyretin mutation, Val30Met. The classification of systemic and ocular amyloidosis as well as the genetics of familial amyloidotic polyneuropathy are briefly reviewed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1742 Prealbumin from human plasma, lyophilized powder