EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A reagent for both visualization and chemiluminescence detection of horseradish peroxidase-tagged DNA probes.
PMID 8600825
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I10201
4-Iodophenol, 99%
C6H5IO