EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Analytical methods for the determination of isoproturon and diflufenican residues in runoff and soil.
PMID 8601057
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45751
Diflufenican, PESTANAL®, analytical standard
C19H11F5N2O2