EMAIL THIS PAGE TO A FRIEND
Nature
Inhibition of G-protein-mediated MAP kinase activation by a new mammalian gene family.
PMID 8602223
Abstract
A general property of signal transduction pathways is that prolonged stimulation decreases responsiveness, a phenomenon termed desensitization. Yeast cells stimulated with mating pheromone activate a heterotrimeric G-protein-linked, MAP-kinase-dependent signalling pathway that induces G1-phase cell-cycle arrest and morphological differentiation (reviewed in refs 1, 2). Eventually the cells desensitize to pheromone and resume growth. Genetic studies have demonstrated the relative importance of a desensitization mechanism that uses the SST2 gene product, Sst2p. Here we identify a mammalian gene family termed RGS (for regulator of G-protein signalling) that encodes structural and functional homologues of Sst2p. Introduction of RGS family members into yeast blunts signal transduction through the pheromone-response pathway. Like SST2 (refs 8-10), they negatively regulate this pathway at a point upstream or at the level of the G protein. The RGS family members also markedly impair MAP kinase activation by mammalian G-protein-linked receptors, indicating the existence and importance of an SST2-like desensitization mechanism in mammalian cells.
Related Materials