EMAIL THIS PAGE TO A FRIEND
FEBS letters
Streptozotocin, an inducer of NAD+ decrease, attenuates M-potassium current inhibition by ATP, bradykinin, angiotensin II, endothelin 1 and acetylcholine in NG108-15 cells.
PMID 8603696
Abstract
The M-potassium current was inhibited by bath application of 100 micron ATP, 10 nM bradykinin, 100 nM angiotensin II and 100 nM endothelin 1 as well as by 10 micron acetylcholine in an m1-muscarinic acetylcholine receptor-transformed NG108-15 cell line. The inhibition of M-current was attenuated in cells pretreated with 5 mM streptozotocin for 5-15 h and restored by simultaneous incubation with 5 mM nicotinamide. The results suggest that signal transduction from these five different receptors to M channels shares a common pathway which is susceptible to a streptozotocin-induced decrease in cellular NAD+ content.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
NG108-15, 88112302