EMAIL THIS PAGE TO A FRIEND
Experimental hematology
Acute inhalation exposure to isobutyl nitrite causes nonspecific blood cell destruction.
PMID 8605963
Abstract
Abuse of nitrite inhalants is widespread among male homosexuals and has been epidemiologically correlated with seropositivity to human immunodeficiency virus (HIV) and to Kaposi's sarcoma. These drugs may act as cofactors in AIDS if they compromise the ability to resist infection or tumor growth. We have previously reported that 14 daily 45-minute exposure to 900 ppm isobutyl nitrite in an inhalation chamber did compromise the immunocompetence of mice. We now report that a single 45-minute exposure produced a transient anemia. Erythrocyte counts, hemoglobin, and hematocrit levels were reduced by 7% but rebounded to above-normal levels 24 hours later. In vitro exposure of blood to isobutyl nitrite vapors did not lyse the cells but did induce Heinz body formation and increase their binding to macrophages. Thus, it is likely that the red cells were removed by phagocytic clearance not by direct lysis. Blood leukocyte numbers were also reduced following a single exposure to the inhalant, but the cell loss was delayed until 24 hours after exposure. Recovery of peripheral blood leukocytes 72 hours after exposure coincided with a reduction in spleen cellularity, suggesting that spleen cells were mobilized to replace lost blood leukocytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
327190
Isobutyl nitrite, 95%
C4H9NO2