EMAIL THIS PAGE TO A FRIEND
Life sciences
Morin hydrate inhibits azo-initiator induced oxidation of human low density lipoprotein.
PMID 8606614
Abstract
We demonstrated that the flavonoid morin hydrate at 75-100 microM protects against the oxidation of low density lipoprotein (LDL) by free radicals produced by 2,2'-azo-bis(2-amidinopropane) dihydrochloride. Morin hydrate reduces the relative electrophoretic mobility, malondialdehyde equivalents and lipid peroxide level of oxidized LDL. On the other hand, Trolox (an analogue of vitamin E) showed less protective effect in the present system. Since free radical mediated oxidation of LDL is implicated to be a cause of atherogenesis, morin hydrate may be a candidate chemotherapeutic agent herein.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M4008
Morin hydrate, powder
C15H10O7 · xH2O