EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Rapid reduction and removal of HDL- but not LDL-associated cholesteryl ester hydroperoxides by rat liver perfused in situ.
PMID 8615764
Abstract
To test whether high-density lipoproteins (HDL) could aid in the removal in vivo of potentially atherogenic oxidized lipids, we perfused rat liver in situ with buffer supplemented with isolated human HDL containing small amounts of cholesteryl linoleate hydro(pero)xides [CH18:2-O(O)H]. Perfusion resulted in the rapid removal of Ch18:2-O(O)H from HDL with a half-life (t1/2)of 11.4 min., faster than that of unoxidized cholesteryl linoleate, and dependent of the presence of the liver. In addition, the liver enhanced the reduction of Ch18:2-OOH associated with HDL remaining in the perfusate buffer. Perfusion resulted in the release of a hepatic activity that enhanced the reduction of HDL-associated CH18:2-OOH and was resistant to heat treatment. In contrast with the situation with HDL, low-density lipoprotein (LDL)-associated CH18:2-O(O)H were neither removed nor reduced by perfused rat liver within the time course studied, in support of a possible role for HDL in the detoxification of circulating lipid hydroperoxides in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0289
Cholesteryl linoleate, ≥98% (HPLC; detection at 205 nm)
C45H76O2