EMAIL THIS PAGE TO A FRIEND
Experimental parasitology
Boophilus microplus: multiple proteolytic activities in the midgut.
PMID 8617328
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6268 Hide–Remazol Brilliant Blue R, protease substrate