EMAIL THIS PAGE TO A FRIEND
Experimental parasitology
Identification of 3-hydroxyproline residues in several proteins of Fasciola hepatica.
PMID 8617333
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
420174
cis-3-Hydroxy-DL-proline, 95%
C5H9NO3