EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Reversible red-ox reactions of the diiron site in the mouse ribonucleotide reductase R2 protein.
PMID 8619810
Abstract
The red-ox reactions of the dinuclear iron center of mouse R2 protein upon interaction with different reductants (dithionite alone and with mediators) and oxidants (PES, methylene blue, hydrogen peroxide and para-benzoquinone) have been studied by EPR and optical spectroscopy. The obtained results indicate that the transitions between Fe(III)Fe(III), Fe(II)Fe(III) and Fe(II)Fe(II) states of the dinuclear iron center are reversible and the mu-oxo-bridge may be formed upon oxidation by non-oxygen oxidants. In contrast to the case for the E. coli R2 protein, dithionite alone reduces the tyrosyl radical and diiron center in mouse R2 protein.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4544
Phenazine ethosulfate, ≥95%
C16H18N2O4S