EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Double tagging recombinant A1- and A2A-adenosine receptors with hexahistidine and the FLAG epitope. Development of an efficient generic protein purification procedure.
PMID 8619901
Abstract
An expression plasmid for mammalian cells (CLDN10B) has been modified to add nucleotides encoding hexahistidine and the FLAG peptide (H/F) to cDNAs. The new mammalian expression plasmid has been named pDoubleTrouble (pDT). The plasmid and a recombinant baculovirus were used to produce native-and H/F-human A1 and A2A adenosine receptors, optimally expressed in CHO-K1 and Sf9 cells, respectively. Binding to recombinant H/F-A1 receptors (Bmax = 30 pmol/mg protein) was characterized using [3H]8-cyclopentyl-1,3-dipropylxanthine ([3H]CPX) and 125I-N6-aminobenzyladenosine (125I-ABA). Binding to H/F-A2A receptors (Bmax = 48 pmol/mg protein) was characterized using [3H]5'-N-ethylcarboxamidoadenosine ([3H]NECA) and [3H]2-[4-(2-carboxyethyl)phenethylamino]-NECA ([3H]CGS21680). By comparison to native receptors, the addition of H/F to the amino termini of these receptors had no effect on the binding affinities cyclic AMP accumulation in intact cells was not affected by the H/F extension. Anti-FLAG and Ni-nitrilotriacetic acid affinity chromatography resulted in high yield ( >50% overall recovery) of nearly homogeneous deglycosylation with N-glycosidase F. We anticipate that pDT will be generally useful for facilitating the purification in high yield of recombinant receptors and other proteins by single or sequential affinity chromatography steps.
Related Materials