EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Cloning of a putative ligand for the T1/ST2 receptor.
PMID 8621446
Abstract
T1/ST2 is a receptor-like molecule homologous to the type I interleukin-1 receptor. Despite this sequence similarity, we have been unable to demonstrate binding of T1/ST2 to any of the three interleukin-1 species. In searching for a ligand for T1/ST2, we have cloned a cell surface protein to which it binds. This protein is unable to initiate signal transduction by the T1/ST2 receptor in several in vitro assays.