EMAIL THIS PAGE TO A FRIEND
Archives of dermatology
Chronic finger dermatitis after trauma. Diagnosis: posttraumatic eczema with allergic contact dermatitis to neomycin, bacitracin, and topical corticosteroids.
PMID 8629854