EMAIL THIS PAGE TO A FRIEND
Life sciences
In vivo pharmacokinetics and in vitro production of cocaethylene in pregnant guinea pigs.
PMID 8637393
Abstract
Simultaneous exposure to cocaine and ethanol results in the formation of cocaethylene, an active metabolite of cocaine. The concurrent abuse of both cocaine and ethanol is common during human pregnancy, but the kinetics of elimination and formation of this ethyl ester of cocaine have not been studied during pregnancy in any species. In the late gestation guinea pig (61 to 63 days), cocaethylene, at doses of 2 to 4 mg.kg-1, is rapidly eliminated with a half-life of 29 min and a total body clearance of 77 ml.min-1.kg-1. It is formed enzymatically by hepatic microsomal preparations from fetal, neonatal and maternal guinea pigs. The maximum rate of cocaethylene production (apparent Vmax) when either ethanol or cocaine are varied while the other substrate is held constant, increases with age, from the late fetal period (65 days gestation, term 70 days) to adulthood. However, the Michaelis-Menten constant (apparent KM) does not change with age. The rapid elimination of cocaethylene, coupled with the slow rate of formation (apparent Vmax of 140 pmol.min-1.mg microsomal protein-1) and the small amount of plasma analyzed most likely explains the inability to detect coacethylene in vivo after concomitant cocaine and ethanol administration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B142
Benzoylnorecgonine, analytical standard
C15H17NO4