EMAIL THIS PAGE TO A FRIEND
Cancer research
Glutathione conjugates of tert-butyl-hydroquinone, a metabolite of the urinary tract tumor promoter 3-tert-butyl-hydroxyanisole, are toxic to kidney and bladder.
PMID 8640754
Abstract
3-tert-Butyl-4-hydroxyanisole and tert-butyl-hydroquinone (TBHQ) are antioxidants known to promote renal and bladder carcinogenesis in the rat, although the mechanisms of these effects are unclear. Because glutathione (GSH) conjugates of a variety of hydroquinones are nephrotoxic, and because 2-tert-butyl-5-(glutathion-S-yl)hydroquinone [5-(GSyl)TBHQ], 2-tert-butyl-6-(glutathion-S-yl)hydroquinone [6-(GSyl)TBHQ], and 2-tert-butyl-3,6-bis-(glutathion-S-yl)hydroquinone [3,6-bis-(GSyl)-TBHQ] have been identified recently as metabolites of TBHQ in the male rat, we investigated the effects of these metabolites in the male rat. At the highest dose tested (400 micromol/kg,i.v.) 5-(Gsyl)TBHQ and 6-(GSyl)TBHQ caused 2-fold increases in the urinary excretion of gamma-glutamyl transpeptidase and alkaline phosphatase, and pigments arising from the polymerization of metabolites were deposited in the kidney. 3,6-bis-(GSyl)TBHQ (200 micromol/kg) was the most potent of the GSH conjugates tested and produced significant increases in the urinary excretion of gamma-glutamyl transpeptidase, alkaline phosphatase, lactate dehydrogenase, and glucose (2-, 2-, 22-, and 11-fold increases, respectively). Alterations in the biochemical parameters correlated with the degree of single cell and tubular necrosis in the S(3)-M segment of the proximal tubule, as observed by light microscopy. In addition to nephrotoxicity, 3,6-bis-(GSyl)TBHQ increased the bladder wet weight 2-fold and caused severe hemorrhaging of the bladder. The half-wave oxidation potentials of 5-(Gsyl)TBHQ and 6-(GSyl)TBHQ were similar to that of TBHQ, whereas the half-wave oxidation potential of 3,6-bis-(Gsyl)TBHQ was approximately 100 mV higher than that of TBHQ. The TBHQ-GSH conjugates also catalyzed the formation of 8- hydroxydeoxyguanosine, indicating that GSH conjugation does not impair the redox activity of TBHQ. Because some chemicals may induce carcinogenesis by a mechanism involving cytotoxicity followed by sustained regenerative hyperplasia, our results suggest that the toxicity of GSH conjugates of TBHQ to kidney and bladder may contribute to the promoting effect of 3-tert-butyl-4-hydroxyanisole and TBHQ in these tissues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
96175
3-tert-Butyl-4-hydroxyanisole, tested according to Ph.Eur.
C11H16O2