EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Design and synthesis of new linear and cyclic bradykinin antagonists.
PMID 8642569
Abstract
We report here on the synthesis and pharmacological properties of a new series of small linear and cyclic peptides derived from the five C-terminal amino acid residues of second-generation bradykinin receptor antagonists. Variations of the two first residues of the pentapeptide (Thi-Ser-D-Tic-Oic-Arg) were shown to modulate the biological activities of the analogs on bradykinin-induced smooth muscle contractions in rabbit jugular vein (RJV), a tissue preparation specific of the B2 bradykinin receptor. Several analogs showed pA2 values around 7 on this tissue preparation, and one cyclic compound, c[-Gly-Thi-D-Tic-Oic-Arg-], 24, in which Thi-Ser was replaced by Gly-Thi, displayed a pA2 of 7.4 on RJV. On the basis of these results, three cyclic molecules and their linear counterparts (compounds 22-24 and 4-6, respectively) were tested on human umbilical vein, a tissue specific of the human B2 receptor. The pKB values obtained for these compounds on these tissue preparations were equivalent to those obtained for the decapeptide NPC 567 (4.8 < pA2 < 5.1). NMR and molecular modeling studies performed on compound 24 clearly demonstrated a type II' beta-turn structure. This analog may serve as a new lead for the design of nonpeptide ligands of the bradykinin B2 receptor subtype.