EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Prostaglandin H synthase 2 is expressed abnormally in human colon cancer: evidence for a transcriptional effect.
PMID 8643486
Abstract
Evidence from epidemiological studies, clinical trials, and animal experiments indicates that inhibitors of prostaglandin synthesis lower the risk of colon cancer. We tested the hypothesis that abnormal expression of prostaglandin H synthase 2 (PHS-2), which can be induced by oncogenes and tumor promoters, occurs during colon carcinogenesis by examining its level in colon tumors. Human colon cancers were found to have an increased expression of PHS-2 mRNA compared with normal colon specimens from the same patient (n = 5). In situ hybridization showed that the neoplastic colonocytes had increased expression of PHS-2 (n = 4). Additionally, five colon cancer cell lines were shown to express high levels of PHS-2 mRNA even in the absence of a known inducer of PHS-2. To study the basis for this increased gene expression, we transfected a colon cancer cell line, HCT-116, with a reporter gene containing 2.0 kb of the 5' regulatory sequence of the PHS-2 gene. Constitutive transcription of the reporter gene was observed, whereas normal control cell lines transcribed the reporter only in response to an exogenous agonist. We conclude that PHS-2 is transcribed abnormally in human colon cancers and that this may be one mechanism by which prostaglandins or related compounds that support carcinogenesis are generated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
HCT 116 Cell Line human, 91091005, colon carcinoma