EMAIL THIS PAGE TO A FRIEND
Mutation research
Reversal of malignancy by the adenovirus E1a gene.
PMID 8657190
Abstract
Tumor suppressor genes such as Rb and p53 usually kill tumor cells when overexpressed ectopically. This is a consequence of their normal cell cycle regulatory functions. By contrast, the E1a gene of adenovirus, a common cold virus, converts tumor cells into viable normal cells. This has advantages for investigation and control of cancer. In particular, E1a is a master programmer of the epithelial phenotype. This provides a new tool for understanding the molecular basis of the epithelial-mesenchymal transition, and how it goes awry in cancer cells. Furthermore, epithelial cells are sensitive to a form of apoptosis - 'anoikis' - that is induced by detachment from extracellular matrix. This property confers strict anchorage-dependence. Transcriptional programming, by E1a or the formation of cell-cell junctional complexes, programs epithelial cells to be sensitive to anoikis.