EMAIL THIS PAGE TO A FRIEND
Calcified tissue international
Inhibition of parathyroid hormone-stimulated resorption in cultured fetal rat long bones by the main metabolites of ipriflavone.
PMID 8661483
Abstract
Ipriflavone is an isoflavone derivative used in the prevention and treatment of postmenopausal and senile osteoporosis in humans. To assess the potential contribution of the main in vivo ipriflavone metabolites (M1, M2, M3, and M5) on the pharmacological properties of the drug, we investigated their effect on osteoclastic resorption induced by the well-known stimulator of bone resorption bovine parathyroid hormone fragment 1-34 (bPTH 1-34). The study was carried out using fetal rat long bones in stationary cultures. The amount of osteoclastic resorption was determined by assaying for 5 days the release from bones in the media of previously incorporated 45Ca. All metabolites were effective at inhibiting osteoclastic resorption. Maximal potency was shown by M3, characterized by a significant effect at 10 microM (P < 0.01) and by an IC50 value of 17 microM. M2 was about threefold less potent than M3 (IC50 = 46 microM). M1 and M5 were the least active compounds with an IC50 value of 117 and 200 microM, respectively. The present evidence indicates that metabolites of ipriflavone, in particular M3 and M2, inhibit bPTH 1-34-induced bone resorption in fetal rat long bones. Accordingly, they may play an important role in the pharmacological effects of the drug.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3671 Parathyroid Hormone Fragment 1-34 bovine, ≥97% (HPLC), powder
C183H288N54O50S2