EMAIL THIS PAGE TO A FRIEND
Pediatric dentistry
A comparison of the pulpal response to freeze-dried bone, calcium hydroxide, and zinc oxide-eugenol in primary teeth in two cynomolgus monkeys.
PMID 8668571
Abstract
This study assessed the pulp healing response to human freeze-dried bone (FDB) in two cynomolgus monkeys using 36 noncarious primary teeth pulpotomized and randomly assigned to three medicaments. FDB was applied on the pulp stumps and covered with sterile tin foil as experimental group. The two other groups received either calcium hydroxide [Ca(OH)2], or IRM (reinforced ZOE). All teeth were restored with amalgam. One animal was sacrificed at 6 weeks and the other 6 months after treatment. Teeth were extracted and placed in 10% formalin. Histological evaluation indicated that 100% of teeth treated with FDB had vital pulps compared with 75% of the Ca(OH)2 group after 6 weeks. Dentin bridges were present in 87.5% of FDB versus 75% of Ca(OH)2 group. Inflammatory cells were absent or mild in 100% of FDB-treated versus 75% of the Ca(OH)2 group. After 6 months, 83.3% of FDB-treated teeth had vital pulps compared with the Ca(OH)2 group, which showed 100% pulpal necrosis. In FDB-treated pulps, 100% of teeth showed dentin bridges versus 50% of teeth treated with Ca(OH)2. Inflammatory cells were absent or mildly present in 83.3% of FDB-treated teeth while 100% of Ca(OH)2 showed moderate to severe inflammation. IRM-treated teeth all showed pulpal necrosis after 6 months. We concluded that FDB was superior to calcium hydroxide in treating primary pulp dentition in cynomolgus monkeys.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF26583194
Tin, foil, 150x150mm, thickness 0.125mm, as rolled, 98.8%
Sn
GF66339254
Tin, foil, 150x150mm, thickness 0.125mm, as rolled, 99.99+%
Sn
GF54760259
Tin, foil, 150x150mm, thickness 0.15mm, as rolled, 98.8%
Sn
GF67327375
Tin, foil, 150x150mm, thickness 0.1mm, as rolled, 98.8%
Sn
GF27720019
Tin, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95%
Sn
GF16685219
Tin, foil, 150x150mm, thickness 0.20mm, as rolled, 98.8%
Sn
GF45567434
Tin, foil, 150x150mm, thickness 0.25mm, as rolled, 99.99+%
Sn
GF28603212
Tin, foil, 150x150mm, thickness 0.5mm, as rolled, 98.8%
Sn
GF28221274
Tin, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99+%
Sn
GF77851323
Tin, foil, 150x150mm, thickness 1.0mm, as rolled, 98.8%
Sn
GF16505636
Tin, foil, 150x150mm, thickness 1.0mm, as rolled, 99.999%
Sn
GF50720925
Tin, foil, 150x150mm, thickness 2.0mm, as rolled, 98.8%
Sn
GF00846413
Tin, foil, 150x150mm, thickness 2.0mm, as rolled, 99.99+%
Sn
GF99151881
Tin, foil, 25x25mm, thickness 0.125mm, as rolled, 98.8%
Sn
GF83193925
Tin, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Sn
GF86850029
Tin, foil, 25x25mm, thickness 0.15mm, as rolled, 98.8%
Sn
GF17353425
Tin, foil, 25x25mm, thickness 0.15mm, as rolled, 99.99+%
Sn
GF40941097
Tin, foil, 25x25mm, thickness 0.1mm, as rolled, 98.8%
Sn
GF66628996
Tin, foil, 4mm disks, thickness 0.007mm, 97.4%
Sn
GF33080532
Tin, foil, 4mm disks, thickness 0.015mm, 99.75%
Sn
GF29177039
Tin, foil, 4mm disks, thickness 0.02mm, 99.75%
Sn
GF51371232
Tin, foil, 4mm disks, thickness 0.125mm, as rolled, 98.8%
Sn
GF34214464
Tin, foil, 4mm disks, thickness 0.25mm, as rolled, 98.8%
Sn
GF63204106
Tin, foil, 4mm disks, thickness 1.0mm, as rolled, 100%
Sn
GF04830331
Tin, foil, 50mm disks, thickness 0.0125mm, 97.4%
Sn
GF96864643
Tin, foil, 50mm disks, thickness 0.05mm, as rolled, 98.8%
Sn
GF07913088
Tin, foil, 8mm disks, thickness 0.06mm, as rolled, 99.99+%
Sn
GF18183764
Tin, foil, 8mm disks, thickness 0.1mm, as rolled, 98.8%
Sn
GF36451098
Tin, foil, 8mm disks, thickness 0.65mm, as rolled, 99.99+%
Sn
GF00022699
Tin, foil, light tested, 100x100mm, thickness 0.0125mm, 97.4%
Sn