EMAIL THIS PAGE TO A FRIEND
Pharmacological research
Endogenous morphine.
PMID 8668642
Abstract
This review surveys the discovery of endogenous alkaloids in mammals. The formation of morphine in mammalian brain was assumed in 1970. The existence of morphine was demonstrated by radioimmunoassay. Identification of morphine was performed by spectroscopic methods. The isolation of mammalian morphine raises the question of biosynthesis. Recently, it has been shown that the biosynthetic pathway is similar to that that exists in poppy.