EMAIL THIS PAGE TO A FRIEND
Voprosy virusologii
[Disinfecting action of chloramine B on Marburg virus].
PMID 8669148
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
23265
N-Chlorobenzenesulfonamide sodium salt, ~28% active chlorine basis
C6H5ClNNaO2S · xH2O