EMAIL THIS PAGE TO A FRIEND
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association
Relative efficacy of haemoperfusion, haemodialysis and CAPD in the removal of procainamide and NAPA in a patient with severe procainamide toxicity.
PMID 8671917
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
269476
N-Acetylprocainamide, ≥99%
C15H23N3O2
A5513
N-Acetylprocainamide hydrochloride, ≥99% (HPLC), powder
C15H23N3O2 · HCl