EMAIL THIS PAGE TO A FRIEND
Farmaco (Societa chimica italiana : 1989)
Synthesis and alpha,beta-adrenergic blocking potency of 2,3-dihydro-1,4-benzodioxin derivatives. (II).
PMID 8688140
Abstract
The four stereoisomers of compound 1 were synthesized from 2,3-dihydro-1, 4-benzodioxin and evaluated as alpha- and beta-adrenergic antagonists. Enantiomer 1-b [2R, 2'S] (Figure 1) is the best beta 1-blocking agent. Furthermore all compounds showed a alpha-blocking activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
179000
Benzo-1,4-dioxane, 97%
C8H8O2