EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
(S)-(-)-4-[4-[2-(isochroman-1-yl)ethyl]-piperazin-1-yl] benzenesulfonamide, a selective dopamine D4 antagonist.
PMID 8691438
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0174
Sonepiprazole hydrate, ≥98% (HPLC)
C21H27N3O3S · xH2O