EMAIL THIS PAGE TO A FRIEND
Planta medica
A competitive enzyme immunoassay for the pyrrolizidine alkaloids of the senecionine type.
PMID 8693043
Abstract
We developed an immunoassay with antibodies against retrorsine to detect the closely related senecionine, the main alkaloid in various Asteraceae. Sensitivity is about 23 pg (68 fmol) for senecionine. Cross reactivity of monocrotalin, retrorsine N-oxide, senkirkine (< 0.1%), and seneciphylline (3.6-34.5%) was resolved. The alkaloid content in leaves of Petasites hybridus (Asteraceae) was determined to be 3.86 ppm, calculated as senecionine, which was far less than in rhizomes (104.8 ppm). Additionally, we proved specificity by taking immunograms from different parts of Petasites hybridus. The test is a suitable tool for assessing the toxic potential of medicinal plants containing senecionine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50351
Senecionine, analytical standard
C18H25NO5
17806
Senecionine, ≥95.0% (GC)
C18H25NO5