EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Inter- and intra-laboratory testing of the Daphnia magna IQ toxicity test.
PMID 8694887
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7633
4-Methylumbelliferyl α-D-galactopyranoside, ≥98% (TLC)
C16H18O8