EMAIL THIS PAGE TO A FRIEND
Journal of vascular research
Does aldosterone-induced cardiac fibrosis involve direct effects on cardiac fibroblasts?
PMID 8695756
Abstract
To investigate the hypothesis that aldosterone plays a role in the development of fibrosis, cultured fibroblasts from adult rat heart have been examined for their expression of aldosterone receptors and the effects of aldosterone on collagen synthesis. Binding assays with both 3H-aldosterone and 3H-RU26752 in intact cardiac fibroblasts and cytosolic extracts from cardiac fibroblasts failed to reveal expression of aldosterone receptors. However, using the method of reverse transcription-polymerase chain reaction, we could demonstrate the expression of mRNA for the mineralocorticoid receptor in both cardiac fibroblasts and neonatal rat cardiomyocytes. Functional studies investigating the effect of aldosterone on collagen synthesis (3H-proline incorporation into collagenous protein) revealed that aldosterone does not stimulate collagen synthesis in cardiac fibroblasts at concentrations (10(-8) to 10(-9) M) observed in primary or secondary hyperaldosteronism. At higher concentrations (10(-6) to 10(-7) M) aldosterone inhibited collagen synthesis. Expression of collagen genes I alpha 1, III alpha 1, IV alpha 1 and of the collagenase gene was not affected by aldosterone. The collagen gene VI alpha 2 was also found to be expressed in cultured cardiac fibroblasts, and its expression was also independent of aldosterone. The data indicate that fibrosis is not due to a direct effect of aldosterone on fibroblast collagen synthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R2153
RU 26752, ≥98% (HPLC), solid
C25H36O3