EMAIL THIS PAGE TO A FRIEND
The Journal of applied bacteriology
The use of an automated assay to assess phage survival after a biocidal treatment.
PMID 8698661
Abstract
The coliphage K1-5 has been used in an automated assay to monitor the viricidal activity of various disinfectants. This indirect assay based on the spectrophotometric reading of the lysis of the host cell (Escherichia coli D837) produced encouraging results and was faster than the overlay counting method (previously studied) which relies on plaque formation. However, differences in sensitivity towards some disinfectants were observed between the two methods.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8986
Alexidine dihydrochloride, ≥95% (HPLC)
C26H56N10 · 2HCl