EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia farmakologiia
[The pharmacokinetics of propranolol and its conjugated metabolites in patients with chronic ischemic heart disease and arterial hypertension with a one-time and course intake of the drug].
PMID 8704616
Abstract
Pharmacokinetics of propranolol (P), 4-hydroxy-propranolol sulfate (4HOP-Sulf), and glucoronides of pharmacologically active S-enantiomer P (S-PG) and ballast R-enantiomer of P (R-PG) in the blood serum of 21 patients with chronic ischemic heart disease and/or arterial hypertension has been studied at a single and course oral P administration. The values od AUC and T1/2 for potentially active 4HOP-Sulf were significantly higher than those for unchanged P at a single and course administration. The values od AUC and T1/2 for for S-PG were approximately three times higher than those for P-PG after both a single and course administration. Thus the results presented show that potentially active 4HOP-Sulf and S-PG (which undergoes a partial deconjugation in an organism at oral administration) may contribute essentially to the value and duration of the P pharmacological effect.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00881
(±)-4-Hydroxypropranolol hydrochloride, analytical standard
C16H21NO3·HCl