EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Erythema-multiforme-like eruption following allergic contact dermatitis from sesquiterpene lactones in herbal medicine.
PMID 8706423
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0415
Alantolactone, ≥98% (HPLC)
C15H20O2